Anti-CYP1B1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-13HEA 2990 DKK
USBIC9095-13H
Anti-CYP1B1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CYP1B1 (cytochrome P450, family 1, subfamily B, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR