Anti-Cytochrome p450 1A1,2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-11UEA 5090 DKK
USBIC9095-11U
Anti-Cytochrome p450 1A1,2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Inhibition/Neutralization Assays: 1-20?l

Type: Primary
Antigen: Cytochrome p450 1A1,2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR