Anti-Cytochrome p450 1A Mouse Monoclonal Antibody [clone: 6D340]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-10ZEA 3320 DKK
USBIC9095-10Z
Anti-Cytochrome p450 1A Mouse Monoclonal Antibody [clone: 6D340]
Antistoffer
Recommended dilution: ELISA: 1:100-1:500; Western Blot: 1:100-1:1000

Type: Primary
Antigen: Cytochrome p450 1A
Clonality: Monoclonal
Clone: 6D340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR