Anti-Cytochrome p450 1A Mouse Monoclonal Antibody [clone: 6D338]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9095-10WEA 3490 DKK
USBIC9095-10W
Anti-Cytochrome p450 1A Mouse Monoclonal Antibody [clone: 6D338]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Western Blot :1:50-1:100

Type: Primary
Antigen: Cytochrome p450 1A
Clonality: Monoclonal
Clone: 6D338
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR