Anti-CYB5R4 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2625]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9094-40-FITCEA 0 DKK
USBIC9094-40-FITC
Anti-CYB5R4 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2625]
Type: Primary
Antigen: CYB5R4 (cytochrome b5 reductase 4)
Clonality: Monoclonal
Clone: 10B2625
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR