Anti-CYBB Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9094-34BEA 3460 DKK
USBIC9094-34B
Anti-CYBB Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: CYBB (cytochrome b-245, beta polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR