Anti-CDADC1 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2170]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9090-08-APEA 0 DKK
USBIC9090-08-AP
Anti-CDADC1 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2170]
Type: Primary
Antigen: CDADC1 (cytidine and dCMP deaminase domain containing 1)
Clonality: Monoclonal
Clone: 10B2170
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR