Anti-CFTR Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9040-06BEA 4180 DKK
USBIC9040-06B
Anti-CFTR Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CFTR (cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR