Anti-CFTR Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9040-01CEA 5040 DKK
USBIC9040-01C
Anti-CFTR Goat polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5-5 ?g/ml

Type: Primary
Antigen: CFTR (cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR