Anti-CFTR/MRP, Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9040-01BEA 2750 DKK
USBIC9040-01B
Anti-CFTR/MRP, Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:128000

Type: Primary
Antigen: CFTR/MRP,
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR