Anti-CRISP-3 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9018-57EA 5280 DKK
USBIC9018-57
Anti-CRISP-3 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:10;

Type: Primary
Antigen: CRISP-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR