Anti-Cysteine-rich, Angiogenic Inducer 61 Rat Monoclonal Antibody [clone: 8H206]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9018-02EA 4320 DKK
USBIC9018-02
Anti-Cysteine-rich, Angiogenic Inducer 61 Rat Monoclonal Antibody [clone: 8H206]
Antistoffer
Recommended dilution: ELISA: 0.5-1 ?g/ml Western Blot: 2?g/ml; Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: Cysteine-rich, Angiogenic Inducer 61
Clonality: Monoclonal
Clone: 8H206
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR