Anti-Cysteine-rich, Angiogenic Inducer 61 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9018-01AEA 3500 DKK
USBIC9018-01A
Anti-Cysteine-rich, Angiogenic Inducer 61 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:100; Western Blot: 1:200

Type: Primary
Antigen: Cysteine-rich, Angiogenic Inducer 61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR