Anti-CSRNP2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9006-10AEA 4120 DKK
USBIC9006-10A
Anti-CSRNP2 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:64000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: CSRNP2 (cysteine-serine-rich nuclear protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR