Anti-Cysteine Conjugate beta Lyase, Cytoplasmic Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9002-02EA 3240 DKK
USBIC9002-02
Anti-Cysteine Conjugate beta Lyase, Cytoplasmic Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Cysteine Conjugate beta Lyase, Cytoplasmic
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR