Anti-CSRP3 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9001-23EA 3460 DKK
USBIC9001-23
Anti-CSRP3 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 01-0. 03?g/ml

Type: Primary
Antigen: CSRP3 (cysteine and glycine-rich protein 3 (cardiac LIM protein))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR