Anti-Cysteine and Glycine-rich Protein 3 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2618]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9001-22C-FITCEA 0 DKK
USBIC9001-22C-FITC
Anti-Cysteine and Glycine-rich Protein 3 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2618]
Type: Primary
Antigen: Cysteine and Glycine-rich Protein 3
Clonality: Monoclonal
Clone: 10B2618
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR