Anti-CSRP2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9001-10DEA 4320 DKK
USBIC9001-10D
Anti-CSRP2 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:64000; Western Blot: 0.1-0. 3?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: CSRP2 (cysteine and glycine-rich protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR