Anti-CSRP2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9001-10BEA 4780 DKK
USBIC9001-10B
Anti-CSRP2 Goat polyclonal antibody
Antistoffer
Recommended dilution: Western Blot:2?g/ml

Type: Primary
Antigen: CSRP2 (cysteine and glycine-rich protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR