Anti-CSRP1 Mouse monoclonal antibody [clone: 2A11]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC9001-09BEA 3500 DKK
USBIC9001-09B
Anti-CSRP1 Mouse monoclonal antibody [clone: 2A11]
Antistoffer
Type: Primary
Antigen: CSRP1 (cysteine and glycine-rich protein 1)
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR