Anti-CYSLTR2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8998-10AEA 4320 DKK
USBIC8998-10A
Anti-CYSLTR2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CYSLTR2 (cysteinyl leukotriene receptor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR