Anti-Nitroso-L-Cysteine Rat Polyclonal Antibody (Unconjugated)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8996-06EA 5030 DKK
USBIC8996-06
Anti-Nitroso-L-Cysteine Rat Polyclonal Antibody (Unconjugated)
Antistoffer
Type: Primary
Antigen: Nitroso-L-Cysteine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR