Anti-CST3 Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8950-29EA 4930 DKK
USBIC8950-29
Anti-CST3 Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1.0 ?g/ml

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR