Anti-CST3 Mouse monoclonal antibody [clone: 11H29]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8950-09AEA 6900 DKK
USBIC8950-09A
Anti-CST3 Mouse monoclonal antibody [clone: 11H29]
Antistoffer
Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Monoclonal
Clone: 11H29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR