Anti-CST3 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8950-05EEA 4120 DKK
USBIC8950-05E
Anti-CST3 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:8000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR