Anti-CST3 Mouse monoclonal antibody [clone: 9L818]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8950-01CEA 9110 DKK
USBIC8950-01C
Anti-CST3 Mouse monoclonal antibody [clone: 9L818]
Antistoffer
Recommended dilution: ELISA: 2-8?g/ml

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Monoclonal
Clone: 9L818
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR