Anti-Cystatin A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8949-19EA 4230 DKK
USBIC8949-19
Anti-Cystatin A Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1:100-1:250

Type: Primary
Antigen: Cystatin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR