Anti-Cystatin A Mouse Monoclonal Antibody [clone: 13A249]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8949-18FEA 3740 DKK
USBIC8949-18F
Anti-Cystatin A Mouse Monoclonal Antibody [clone: 13A249]
Antistoffer
Recommended dilution: Western Blot: 1-4?g/ml

Type: Primary
Antigen: Cystatin A
Clonality: Monoclonal
Clone: 13A249
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR