Anti-CBS Mouse Monoclonal Antibody [clone: 5F7]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC8948-50GEA 5250 DKK
USBIC8948-50G
Anti-CBS Mouse Monoclonal Antibody [clone: 5F7]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Monoclonal
Clone: 5F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human