Anti-CBS Mouse Monoclonal Antibody [clone: 6A9]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8948-50FEA 4790 DKK
USBIC8948-50F
Anti-CBS Mouse Monoclonal Antibody [clone: 6A9]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Monoclonal
Clone: 6A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR