Anti-CBS Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8948-50C-FITCEA 4620 DKK
USBIC8948-50C-FITC
Anti-CBS Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR