Anti-Cystathionase Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2622]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8948-15-PEEA 0 DKK
USBIC8948-15-PE
Anti-Cystathionase Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2622]
Type: Primary
Antigen: Cystathionase
Clonality: Monoclonal
Clone: 10B2622
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR