Anti-CYP7B1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8929-05A2EA 3460 DKK
USBIC8929-05A2
Anti-CYP7B1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Immunochemistry: 3-6?g/ml

Type: Primary
Antigen: CYP7B1 (cytochrome P450, family 7, subfamily B, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR