Anti-CYP3A7 Mouse Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8929-04CEA 3500 DKK
USBIC8929-04C
Anti-CYP3A7 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP3A7 (cytochrome P450, family 3, subfamily A, polypeptide 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR