Anti-CYP26B1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8929-03H1EA 4320 DKK
USBIC8929-03H1
Anti-CYP26B1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:3000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CYP26B1 (cytochrome P450, family 26, subfamily B, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR