Anti-Cylicin I Mouse Monoclonal Antibody [clone: 5F176]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8925-26EA 3230 DKK
USBIC8925-26
Anti-Cylicin I Mouse Monoclonal Antibody [clone: 5F176]
Antistoffer
Type: Primary
Antigen: Cylicin I
Clonality: Monoclonal
Clone: 5F176
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR