Anti-Cylicin I Mouse Monoclonal Antibody [clone: 5F175]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8925-25EA 3530 DKK
USBIC8925-25
Anti-Cylicin I Mouse Monoclonal Antibody [clone: 5F175]
Antistoffer
Type: Primary
Antigen: Cylicin I
Clonality: Monoclonal
Clone: 5F175
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR