Anti-CYLD Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8923-50DEA 3460 DKK
USBIC8923-50D
Anti-CYLD Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: CYLD (cylindromatosis (turban tumor syndrome))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR