Anti-CYLD Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8923-50AEA 3310 DKK
USBIC8923-50A
Anti-CYLD Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYLD (cylindromatosis (turban tumor syndrome))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR