Anti-CYFIP1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8920-25EA 2750 DKK
USBIC8920-25
Anti-CYFIP1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1: 2000

Type: Primary
Antigen: CYFIP1 (cytoplasmic FMR1 interacting protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR