Anti-Cyclophilin G Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2629]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8855-70-APCEA 0 DKK
USBIC8855-70-APC
Anti-Cyclophilin G Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2629]
Type: Primary
Antigen: Cyclophilin G
Clonality: Monoclonal
Clone: 10B2629
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR