Anti-PPIE Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8855-46EA 4700 DKK
USBIC8855-46
Anti-PPIE Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: PPIE (peptidylprolyl isomerase E (cyclophilin E))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR