Anti-PPID Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8855-20BEA 3460 DKK
USBIC8855-20B
Anti-PPID Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:64000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: PPID (peptidylprolyl isomerase D)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR