Anti-PPID Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8855-20EA 5420 DKK
USBIC8855-20
Anti-PPID Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: PPID (peptidylprolyl isomerase D)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Hamster, Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR