Anti-PPIB Mouse Monoclonal Antibody [clone: 9H108]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8855-01BEA 4500 DKK
USBIC8855-01B
Anti-PPIB Mouse Monoclonal Antibody [clone: 9H108]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: PPIB (peptidylprolyl isomerase B (cyclophilin B))
Clonality: Monoclonal
Clone: 9H108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR