Anti-PPIA Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8850-03EA 10000 DKK
USBIC8850-03
Anti-PPIA Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1:300

Type: Primary
Antigen: PPIA (peptidylprolyl isomerase A (cyclophilin A))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR