Anti-CDKN3 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8471-67EA 3810 DKK
USBIC8471-67
Anti-CDKN3 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CDKN3 (cyclin-dependent kinase inhibitor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR