Anti-CDKN2A Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8471-65NEA 4730 DKK
USBIC8471-65N
Anti-CDKN2A Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 0.1-1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR