Anti-CCNT1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8462-01EA 4730 DKK
USBIC8462-01
Anti-CCNT1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:1000-1:5000; Western Blot: 1:500-1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: CCNT1 (cyclin T1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR