Anti-CCNL2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8459-60EA 4730 DKK
USBIC8459-60
Anti-CCNL2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:3000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CCNL2 (cyclin L2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR